accessibility menu, dialog, popup

UserName

Viewing profile as user:
HON

CAS RN 874-89-5

IUPAC Name: 4-(hydroxymethyl)benzonitrile
Synonyms: 4-(hydroxymethyl)benzonitrile
Molecular Weight (g/mol): 133.15
Molecular Formula: C8H7NO
InChi Key: XAASLEJRGFPHEV-UHFFFAOYSA-N
SMILES: OCC1=CC=C(C=C1)C#N
Percent Purity 
  • (1)
  • (6)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (6)
  • (1)
  • (6)
  • (3)
  • (1)
  • (1)
  • (2)