accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 874-98-6

IUPAC Name: 1-(bromomethyl)-3-methoxybenzene
Synonyms: 1-(bromomethyl)-3-methoxybenzene
Molecular Weight (g/mol): 201.06
Molecular Formula: C8H9BrO
InChi Key: ZKSOJQDNSNJIQW-UHFFFAOYSA-N
SMILES: COC1=CC=CC(CBr)=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)