accessibility menu, dialog, popup

UserName

Viewing profile as user:
BONHOHFOHHO

CAS RN 874289-21-1

IUPAC Name: {3-fluoro-4-[(2-hydroxyethyl)carbamoyl]phenyl}boronic acid
Synonyms: 3-fluoro-4-[(2-hydroxyethyl)carbamoyl]phenylboronic acid
Molecular Weight (g/mol): 227
Molecular Formula: C9H11BFNO4
InChi Key: RNKVHZGEFMLELU-UHFFFAOYSA-N
SMILES: OCCNC(=O)C1=C(F)C=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results