accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 874289-39-1

IUPAC Name: (5-carbamoyl-2-fluorophenyl)boronic acid
Synonyms: 5-carbamoyl-2-fluorophenylboronic acid
Molecular Weight (g/mol): 182.95
Molecular Formula: C7H7BFNO3
InChi Key: NDVPMVJEURRBQM-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC(B(O)O)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results