accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 874473-14-0

IUPAC Name: 3-methyloxetan-3-amine
Synonyms: 3-methyloxetan-3-amine
Molecular Weight (g/mol): 87.12
Molecular Formula: C4H9NO
InChi Key: NQVWMPOQWBDSAI-UHFFFAOYSA-N
SMILES: CC1(N)COC1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)