accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNICl

CAS RN 874676-81-0

IUPAC Name: 5-chloro-2-iodopyrimidine
Synonyms: 5-chloro-2-iodopyrimidine
Molecular Weight (g/mol): 240.43
Molecular Formula: C4H2ClIN2
InChi Key: BWDLBDLGVMLSCS-UHFFFAOYSA-N
SMILES: ClC1=CN=C(I)N=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)