CAS RN 874781-09-6
IUPAC Name:
4-(difluoromethoxy)benzene-1-sulfonamide
Synonyms:
4-(difluoromethoxy)benzenesulfonamide
Molecular Weight (g/mol):
223.19
Molecular Formula:
C7H7F2NO3S
InChi Key:
BBBQHNZWCPUJNW-UHFFFAOYSA-N
SMILES:
NS(=O)(=O)C1=CC=C(OC(F)F)C=C1
1
–
1
of
1
results
1
4-(Difluoromethoxy)benzenesulfonamide 98.0+%, TCI America™
CAS: 874781-09-6 Molecular Formula: C7H7F2NO3S Molecular Weight (g/mol): 223.19 MDL Number: MFCD03407973 InChI Key: BBBQHNZWCPUJNW-UHFFFAOYSA-N PubChem CID: 24820749 IUPAC Name: 4-(difluoromethoxy)benzene-1-sulfonamide SMILES: NS(=O)(=O)C1=CC=C(OC(F)F)C=C1