accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrH2NFFFF

CAS RN 875664-27-0

IUPAC Name: 2-bromo-4-fluoro-6-(trifluoromethyl)aniline
Synonyms: 2-bromo-4-fluoro-6-(trifluoromethyl)aniline
Molecular Weight (g/mol): 258.01
Molecular Formula: C7H4BrF4N
InChi Key: OTYCOVZULSPPPW-UHFFFAOYSA-N
SMILES: NC1=C(Br)C=C(F)C=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results