accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiCH3CH3NNN

CAS RN 87576-94-1

IUPAC Name: (azidomethyl)trimethylsilane
Synonyms: (azidomethyl)trimethylsilane
Molecular Weight (g/mol): 129.24
Molecular Formula: C4H11N3Si
InChi Key: HKBUFTCADGLKAS-UHFFFAOYSA-N
SMILES: C[Si](C)(C)CN=[N+]=[N-]
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results