accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHH3C

CAS RN 876-02-8

IUPAC Name: 1-(4-hydroxy-3-methylphenyl)ethan-1-one
Synonyms: 1-(4-hydroxy-3-methylphenyl)ethanone
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: LXBHHIZIQVZGFN-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(O)C(C)=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (3)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)