accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 876161-05-6

IUPAC Name: 5-bromopyrazine-2-carboxylic acid
Synonyms: 5-bromopyrazine-2-carboxylic acid
Molecular Weight (g/mol): 203
Molecular Formula: C5H3BrN2O2
InChi Key: NPJHCKULJAIWGV-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=C(Br)C=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results