accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 87617-29-6

IUPAC Name: 4-(propan-2-yl)-2-(trifluoromethyl)aniline
Synonyms: 4-isopropyl-2-(trifluoromethyl)aniline
Molecular Weight (g/mol): 203.21
Molecular Formula: C10H12F3N
InChi Key: ZFHAHXSEZUPPNM-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(N)C(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results