accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 876316-30-2

IUPAC Name: 3-[(1H-1,2,4-triazol-1-yl)methyl]benzaldehyde
Synonyms: 3-(1,2,4-triazol-1-ylmethyl)benzaldehyde
Molecular Weight (g/mol): 187.2
Molecular Formula: C10H9N3O
InChi Key: IBVURYHFXBPNCW-UHFFFAOYSA-N
SMILES: O=CC1=CC(CN2C=NC=N2)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results