accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 876316-43-7

IUPAC Name: 4-(4-isocyanatopyridin-2-yl)morpholine
Synonyms: 4-(4-isocyanatopyridin-2-yl)morpholine
Molecular Weight (g/mol): 205.22
Molecular Formula: C10H11N3O2
InChi Key: LRVCWTODNVVIOK-UHFFFAOYSA-N
SMILES: O=C=NC1=CC(=NC=C1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results