accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNSCl

CAS RN 876316-96-0

IUPAC Name: 5-(chloromethyl)-1-methyl-3-(thiophen-2-yl)-1H-pyrazole
Synonyms: 5-(chloromethyl)-1-methyl-3-(thiophen-2-yl)pyrazole
Molecular Weight (g/mol): 212.7
Molecular Formula: C9H9ClN2S
InChi Key: GQMRXNSYTBKJTB-UHFFFAOYSA-N
SMILES: CN1N=C(C=C1CCl)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results