accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 87666-86-2

IUPAC Name: 3-iodo-5-phenyl-1,1'-biphenyl
Synonyms: 3-iodo-5-phenyl-1,1'-biphenyl
Molecular Weight (g/mol): 356.21
Molecular Formula: C18H13I
InChi Key: LICOFADCKUKHJI-UHFFFAOYSA-N
SMILES: IC1=CC(=CC(=C1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results