accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 876728-39-1

IUPAC Name: 1-(1-methyl-3-phenyl-1H-pyrazol-5-yl)methanamine
Synonyms: 1-(2-methyl-5-phenylpyrazol-3-yl)methanamine
Molecular Weight (g/mol): 187.25
Molecular Formula: C11H13N3
InChi Key: XAVNJYHSYZUYFI-UHFFFAOYSA-N
SMILES: CN1N=C(C=C1CN)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results