accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)NNHOCH3OOOH3CCH3CH3CH3CH3

CAS RN 87694-52-8

IUPAC Name: tert-butyl N-[(1S)-1-[methoxy(methyl)carbamoyl]-2-methylpropyl]carbamate
Synonyms: tert-butyl N-[(1S)-1-[methoxy(methyl)carbamoyl]-2-methylpropyl]carbamate
Molecular Weight (g/mol): 260.33
Molecular Formula: C12H24N2O4
InChi Key: RRBFCGUIFHFYQK-VIFPVBQESA-N
SMILES: CON(C)C(=O)[C@@H](NC(=O)OC(C)(C)C)C(C)C
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)