accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrHNO

CAS RN 877-03-2

IUPAC Name: 5-bromo-1H-indole-3-carbaldehyde
Synonyms: 5-bromo-1H-indole-3-carbaldehyde
Molecular Weight (g/mol): 224.06
Molecular Formula: C9H6BrNO
InChi Key: PEENKJZANBYXNB-UHFFFAOYSA-N
SMILES: BrC1=CC=C2NC=C(C=O)C2=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)