accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 877-09-8

IUPAC Name: 1,2,3,5-tetrachloro-4,6-dimethylbenzene
Synonyms: tetrachloro-m-xylene
Molecular Weight (g/mol): 243.94
Molecular Formula: C8H6Cl4
InChi Key: NTUBJKOTTSFEEV-UHFFFAOYSA-N
SMILES: CC1=C(Cl)C(Cl)=C(Cl)C(C)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results