accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 877-11-2

IUPAC Name: 1,2,3,4,5-pentachloro-6-methylbenzene
Synonyms: pentachlorotoluene
Molecular Weight (g/mol): 264.35
Molecular Formula: C7H3Cl5
InChi Key: AVSIMRGRHWKCAY-UHFFFAOYSA-N
SMILES: CC1=C(Cl)C(Cl)=C(Cl)C(Cl)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results