accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3

CAS RN 877-43-0

IUPAC Name: 2,6-dimethylquinoline
Synonyms: 2,6-dimethylquinoline
Molecular Weight (g/mol): 157.22
Molecular Formula: C11H11N
InChi Key: JJPSZKIOGBRMHK-UHFFFAOYSA-N
SMILES: CC1=CC=C2N=C(C)C=CC2=C1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)