CAS RN 87700-67-2
IUPAC Name:
4-methylnaphthalene-1-carbonyl chloride
Synonyms:
4-methylnaphthalene-1-carbonyl chloride
Molecular Weight (g/mol):
204.65
Molecular Formula:
C12H9ClO
InChi Key:
IXGMHYHWSPDNNS-UHFFFAOYSA-N
SMILES:
CC1=C2C=CC=CC2=C(C=C1)C(Cl)=O
1
–
1
of
1
results
1
4-Methyl-1-naphthoyl chloride, 98%, Thermo Scientific™
CAS: 87700-67-2 Molecular Formula: C12H9ClO Molecular Weight (g/mol): 204.653 MDL Number: MFCD12025362 InChI Key: IXGMHYHWSPDNNS-UHFFFAOYSA-N Synonym: 1-naphthalenecarbonylchloride, 4-methyl,1-naphthalenecarbonyl chloride, 4-methyl,4-methyl-1-naphthalenecarbonyl chloride,1-naphthalenecarbonyl chloride, 4-methyl-9ci,4-methyl-1-napthoyl chloride,4-methyl-1-naphthoyl chloride,4-methyl-naphthalene-1-carbonyl chloride PubChem CID: 24887412 IUPAC Name: 4-methylnaphthalene-1-carbonyl chloride SMILES: CC1=CC=C(C2=CC=CC=C12)C(=O)Cl