accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3SiH3CCH3OOFFF

CAS RN 87736-75-2

IUPAC Name: 1-(4-{[(tert-butyldimethylsilyl)oxy]methyl}phenyl)-2,2,2-trifluoroethan-1-one
Synonyms: 1-(4-{[(tert-butyldimethylsilyl)oxy]methyl}phenyl)-2,2,2-trifluoroethanone
Molecular Weight (g/mol): 318.41
Molecular Formula: C15H21F3O2Si
InChi Key: PNTKHPCHUVZRCB-UHFFFAOYSA-N
SMILES: CC(C)(C)[Si](C)(C)OCC1=CC=C(C=C1)C(=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results