accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNNFFF

CAS RN 87736-88-7

IUPAC Name: {4-[3-(trifluoromethyl)-3H-diazirin-3-yl]phenyl}methanol
Synonyms: {4-[3-(trifluoromethyl)diazirin-3-yl]phenyl}methanol
Molecular Weight (g/mol): 216.16
Molecular Formula: C9H7F3N2O
InChi Key: UZHHTEASLDJYBO-UHFFFAOYSA-N
SMILES: OCC1=CC=C(C=C1)C1(N=N1)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results