accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CClFClOH

CAS RN 877397-65-4

IUPAC Name: 1-(2,6-dichloro-3-fluorophenyl)ethan-1-ol
Synonyms: 1-(2,6-dichloro-3-fluorophenyl)ethanol
Molecular Weight (g/mol): 209.04
Molecular Formula: C8H7Cl2FO
InChi Key: JAOYKRSASYNDGH-UHFFFAOYNA-N
SMILES: CC(O)C1=C(Cl)C=CC(F)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results