accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 877399-74-1

IUPAC Name: tert-butyl 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl]piperidine-1-carboxylate
Synonyms: tert-butyl 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazol-1-yl]piperidine-1-carboxylate
Molecular Weight (g/mol): 377.29
Molecular Formula: C19H32BN3O4
InChi Key: QSQWENQPOSRWLP-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC(CC1)N1C=C(C=N1)B1OC(C)(C)C(C)(C)O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results