accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 87769-39-9

IUPAC Name: tetramethyl-2,4-dihydro-1,3-dioxin-4-one
Synonyms: tetramethyl-1,3-dioxin-4-one
Molecular Weight (g/mol): 156.18
Molecular Formula: C8H12O3
InChi Key: NZEPEQVPVAXLSY-UHFFFAOYSA-N
SMILES: CC1=C(C)C(=O)OC(C)(C)O1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results