accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3H3CSiCH3CH3H3CH3C

CAS RN 87778-95-8

IUPAC Name: trimethyl(1,2,3,4,5-pentamethylcyclopenta-2,4-dien-1-yl)silane
Synonyms: trimethyl(1,2,3,4,5-pentamethylcyclopenta-2,4-dien-1-yl)silane
Molecular Weight (g/mol): 208.42
Molecular Formula: C13H24Si
InChi Key: WNTWQEUDFDAMBF-UHFFFAOYSA-N
SMILES: CC1=C(C)C(C)(C(C)=C1C)[Si](C)(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results