accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOO

CAS RN 878-00-2

IUPAC Name: 4-formylphenyl acetate
Synonyms: 4-formylphenyl acetate
Molecular Weight (g/mol): 164.16
Molecular Formula: C9H8O3
InChi Key: SEVSMVUOKAMPDO-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=C(C=O)C=C1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)