accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH

CAS RN 878-11-5

IUPAC Name: 3-heptyl-1H-pyrrole
Synonyms: 3-heptyl-1H-pyrrole
Molecular Weight (g/mol): 165.28
Molecular Formula: C11H19N
InChi Key: OVBAZQHUSHSARW-UHFFFAOYSA-N
SMILES: CCCCCCCC1=CNC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results