accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CNN

CAS RN 878194-91-3

IUPAC Name: 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-4-carbonitrile
Synonyms: 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-4-carbonitrile
Molecular Weight (g/mol): 230.07
Molecular Formula: C12H15BN2O2
InChi Key: DQQFRFWEPQBNEN-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C(C=CN=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results