accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 87820-88-0

IUPAC Name: 2-[1-(ethoxyamino)propylidene]-5-(2,4,6-trimethylphenyl)cyclohexane-1,3-dione
Synonyms: 2-[1-(ethoxyamino)propylidene]-5-(2,4,6-trimethylphenyl)cyclohexane-1,3-dione
Molecular Weight (g/mol): 329.44
Molecular Formula: C20H27NO3
InChi Key: SOTLWPHEAQOHHC-UHFFFAOYSA-N
SMILES: CCONC(CC)=C1C(=O)CC(CC1=O)C1=C(C)C=C(C)C=C1C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results