accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOHOOOONNH(S)CH3CH3(S)CH3CH3H3C

CAS RN 878797-09-2

IUPAC Name: (4S)-3-[(2S)-3-[4-(tert-butoxy)phenyl]-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoyl]-2,2-dimethyl-1,3-oxazolidine-4-carboxylic acid
Synonyms: (4S)-3-[(2S)-3-[4-(tert-butoxy)phenyl]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoyl]-2,2-dimethyl-1,3-oxazolidine-4-carboxylic acid
Molecular Weight (g/mol): 586.69
Molecular Formula: C34H38N2O7
InChi Key: DNJYPUYQFPFOKS-VMPREFPWSA-N
SMILES: CC(C)(C)OC1=CC=C(C[C@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(=O)N2[C@@H](COC2(C)C)C(O)=O)C=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)