accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NOHN

CAS RN 879-37-8

IUPAC Name: 2-(1H-indol-3-yl)acetamide
Synonyms: 2-(1H-indol-3-yl)acetamide
Molecular Weight (g/mol): 174.2
Molecular Formula: C10H10N2O
InChi Key: ZOAMBXDOGPRZLP-UHFFFAOYSA-N
SMILES: NC(=O)CC1=CNC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results