accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 879-39-0

IUPAC Name: 1,2,3,4-tetrachloro-5-nitrobenzene
Synonyms: 1,2,3,4-tetrachloro-5-nitrobenzene
Molecular Weight (g/mol): 260.88
Molecular Formula: C6HCl4NO2
InChi Key: MTBYTWZDRVOMBR-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Cl)=C(Cl)C(Cl)=C1Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results