accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 879-65-2

IUPAC Name: quinoxaline-2-carboxylic acid
Synonyms: quinoxaline-2-carboxylic acid
Molecular Weight (g/mol): 174.16
Molecular Formula: C9H6N2O2
InChi Key: UPUZGXILYFKSGE-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=C2C=CC=CC2=N1
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)