accessibility menu, dialog, popup

UserName

Viewing profile as user:
FClFO

CAS RN 879093-02-4

IUPAC Name: 4-chloro-2,5-difluorobenzaldehyde
Synonyms: 4-chloro-2,5-difluorobenzaldehyde
Molecular Weight (g/mol): 176.55
Molecular Formula: C7H3ClF2O
InChi Key: XPZYVMBPIANKJG-UHFFFAOYSA-N
SMILES: FC1=CC(Cl)=C(F)C=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results