accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2CH2CCH2

CAS RN 87924-01-4

IUPAC Name: 1,2-diethenylbenzene; ethenylbenzene
Synonyms: divinylbenzene; styrene
Molecular Weight (g/mol): 234.34
Molecular Formula: C18H18
InChi Key: CHRJZRDFSQHIFI-UHFFFAOYSA-N
SMILES: C=CC1=CC=CC=C1.C=CC1=CC=CC=C1C=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)