accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CMgBr

CAS RN 87942-08-3

IUPAC Name: bromo(4-propylphenyl)magnesium
Synonyms: bromo(4-propylphenyl)magnesium
Molecular Weight (g/mol): 223.4
Molecular Formula: C9H11BrMg
InChi Key: HFPDWJMICZDKNO-UHFFFAOYSA-M
SMILES: CCCC1=CC=C([Mg]Br)C=C1
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results