accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 879643-71-7

IUPAC Name: methanimidamide hydroiodide
Synonyms: formamidine hydroiodide
Molecular Weight (g/mol): 171.97
Molecular Formula: CH5IN2
InChi Key: QHJPGANWSLEMTI-UHFFFAOYSA-N
SMILES: I.NC=N
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)