CAS RN 879894-27-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
CERILLIANT™ 7-Aminoflunitrazepam-d7 Solution, 100 μg/mL in acetonitrile, Sold by MilliporeSigma™ Supelco™
This stable-labeled internal standard is suitable for quantitation of 7-aminoflunitrazepam levels in GC/MS or LC/MS methods for clinical toxicology, forensic analysis, urine drug testing, pain prescription monitoring or isotope dilution applications.