accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 879896-48-7

IUPAC Name: 1-methyl-3-(thiophen-2-yl)-1H-pyrazole-5-carbaldehyde
Synonyms: 2-methyl-5-(thiophen-2-yl)pyrazole-3-carbaldehyde
Molecular Weight (g/mol): 192.24
Molecular Formula: C9H8N2OS
InChi Key: RKARRIJZFBGKSH-UHFFFAOYSA-N
SMILES: CN1N=C(C=C1C=O)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results