accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 879896-58-9

IUPAC Name: methyl({[2-(piperidin-1-yl)pyridin-4-yl]methyl})amine
Synonyms: methyl({[2-(piperidin-1-yl)pyridin-4-yl]methyl})amine
Molecular Weight (g/mol): 205.31
Molecular Formula: C12H19N3
InChi Key: CPEUGLUNQVAGDV-UHFFFAOYSA-N
SMILES: CNCC1=CC(=NC=C1)N1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results