accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 88-22-2

IUPAC Name: 2-amino-3,5-dimethylbenzene-1-sulfonic acid
Synonyms: 2-amino-3,5-dimethylbenzenesulfonic acid
Molecular Weight (g/mol): 201.24
Molecular Formula: C8H11NO3S
InChi Key: CFCXQQUQLZIZPI-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(N)C(=C1)S(O)(=O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results