accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 88-58-4

IUPAC Name: 2,5-di-tert-butylbenzene-1,4-diol
Synonyms: di-T-butylhydroquinone
Molecular Weight (g/mol): 222.33
Molecular Formula: C14H22O2
InChi Key: JZODKRWQWUWGCD-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(O)=C(C=C1O)C(C)(C)C
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (2)
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (4)
  • (2)
  • (6)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (4)