accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HOCH3H3CCH3

CAS RN 88-60-8

IUPAC Name: 2-tert-butyl-5-methylphenol
Synonyms: 2-tert-butyl-5-methylphenol
Molecular Weight (g/mol): 164.25
Molecular Formula: C11H16O
InChi Key: XOUQAVYLRNOXDO-UHFFFAOYSA-N
SMILES: CC1=CC=C(C(O)=C1)C(C)(C)C
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)