accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CSOHOO

CAS RN 88-61-9

IUPAC Name: 2,4-dimethylbenzene-1-sulfonic acid
Synonyms: 2,4-dimethylbenzenesulfonic acid
Molecular Weight (g/mol): 186.23
Molecular Formula: C8H10O3S
InChi Key: CHZLVSBMXZSPNN-UHFFFAOYSA-N
SMILES: CC1=CC=C(C(C)=C1)S(O)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results