accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 88-73-3

IUPAC Name: 1-chloro-2-nitrobenzene
Synonyms: o-chloronitrobenzene
Molecular Weight (g/mol): 157.55
Molecular Formula: C6H4ClNO2
InChi Key: BFCFYVKQTRLZHA-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1Cl
Boiling Point 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (2)
  • (5)
  • (3)
  • (4)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)