accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CS

CAS RN 880-36-4

IUPAC Name: 2-octylthiophene
Synonyms: 2-octylthiophene
Molecular Weight (g/mol): 196.35
Molecular Formula: C12H20S
InChi Key: GIFWAJGKWIDXMY-UHFFFAOYSA-N
SMILES: CCCCCCCCC1=CC=CS1
Boiling Point 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)